Learn More
Specifications
Specifications
| Type | Controlled Environments Apron |
| Material | Polyethylene |
| Color | White |
| Thickness | 1 mil |
| For Use With (Application) | Bio and Pharmaceutical Production, Food Processing, Controlled Environments |
| Dimensions (L x W) | 28 x 46 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.