Learn More
Apexbio Technology LLC (-)-epicatechin gallate,5mg CAS# 1257-08-5
SDP
Supplier:  Apexbio Technology LLC B49655
Encompass_Preferred
(-)-epicatechin gallate is a major catechin component in green tea [1]. (-)-epicatechin gallate (ECG) plays an important role in cell growth inhibition, apoptosis and membrane transport system [1]. (-)-epicatechin gallate is a kind of catechin. In HCT-116 cells, ECG activated transcription factor 3 (ATF3), which played a critical role in pro-apoptosis. EGR-1 was involved in ECG-induced ATF3 expression. In HCT-116 cells, ECG (50 µM) increased NAG-1 and ATF3 expression in time- and dose-dependent way [1]. In carcinoma HSC-2 cells, ECG (50 µM) exhibited cytotoxicity with midpoint cytotoxicity (NR50) value of 67 µM. However, in normal HGF-2 fibroblasts, ECG exhibited cytotoxicity at concentrations up to 25 µM with NR50 value of 100 µM. In carcinoma HSC-2 cells, ECG (250 µM) induced nucleosomal DNA fragmentation and apoptosis. ECG (150 µM) significantly increased caspase-3 activity [2]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0361
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.