Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Epicatechin

Catalog No. aaj61218fpl
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61218FPL 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61218FPL Supplier Thermo Scientific Chemicals Supplier No. J61218FPL
Only null left
Add to Cart

CAS: 490-46-0 | C15H14O6 | 290.271 g/mol

An antioxidant and natural product from green tea

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-46-0
Molecular Formula C15H14O6
Molecular Weight (g/mol) 290.271
MDL Number MFCD00075648
InChI Key PFTAWBLQPZVEMU-UKRRQHHQSA-N
Synonym --epicatechin, epicatechin, l-epicatechin, --epicatechol, l-acacatechin, epicatechol, l-epicatechol, 2r,3r-2-3,4-dihydroxyphenyl chroman-3,5,7-triol, -epicatechin, 2r,3r---epicatechin
PubChem CID 72276
ChEBI CHEBI:90
IUPAC Name (2R,3R)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)O

Specifications

Melting Point 240°dec
Color White to Yellow
Quantity 20 mg
Beilstein 92760
Solubility Information Soluble in water or alcohol
Formula Weight 290.27
Physical Form Crystalline Powder
Chemical Name or Material (-)-Epicatechin
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 207-710-1
RTECSNumber KB3745000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.