Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epidermal Growth Factor Receptor (EGFR) Rabbit Monoclonal Antibody, Epredia™

Catalog No. p-4541934 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
7 mL
12 mL
Form:
Ready-to-Use
Supernatant
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
RM2111R1 1 mL Ready-to-Use
RM2111R7 7 mL Ready-to-Use
RM2111RQ 12 mL Ready-to-Use
RM2111S 1 mL Supernatant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. RM2111R1 Supplier Epredia Supplier No. RM2111R1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ EGFR, Rabbit Monoclonal Antibody.

EGFR is a type I receptor tyrosine kinase with sequence homology to c-erbB1, B2, B3, B4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-alpha (TGF-alpha), Heparin-binding EGF (HB-EGF), amphiregulin, betacellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, colon, lung, stomach, head and neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Rabbit

Clone: EP38Y

Species Reactivity: Human. Others not tested.

Ig Isotype: Rabbit IgG

Epitope: Not determined

Immunogen: A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR

Molecular Weight: 175 kDa

Positive Control: Squamous carcinoma

Cellular Localization: Cell membrane and occasional cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen anti-EGFR
Applications Immunohistochemistry
Classification Monoclonal
Clone EP38Y
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Rabbit
Immunogen A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR
Quantity 1 mL
Primary or Secondary Primary
Target Species Human
Form Ready-to-Use
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.