Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epigallocatechin Gallate, G-Biosciences
SCI_ED

Catalog No. S139845
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139845 100 mg
S139727 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139845 Supplier G-Biosciences Supplier No. RC1009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Epigallocatechin gallate (EGCG) has been shown to have a broad spectrum of antibacterial activity against gram positive and some gram negative bacteria.

It is used synergistically with penicillin binding protein inhibitors because both target peptidoglycan on the cell wall.

Chemical Identifiers

CAS 989-51-5
Molecular Weight (g/mol) 458.37 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Ion Chromatography, Mass Spectrometry
Certifications/Compliance ISO, ISO 9001, REACH, GHS/OSHA, NIST
Type Multi-component Standard
Element Multi-element
Industry Type Biotech
Quantity 100 mg
Percent Purity 98%
Grade Laboratory
Chemical Name or Material Epigallocatechin gallate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.