CAS RN 989-51-5
Thermo Scientific Chemicals (-)-Epigallocatechin gallate, 95%
CAS: 989-51-5 Molecular Formula: C22H18O11 Molecular Weight (g/mol): 458.37 MDL Number: MFCD00075940 InChI Key: WMBWREPUVVBILR-WIYYLYMNSA-N Synonym: =--epigallocatechin gallate,egcg,epigallocatechin gallate,epigallocatechin 3-gallate,tea catechin,epigallocatechin-3-gallate,=--epigallocatechin-3-o-gallate,teavigo,=--epigallocatechol gallate,epigallocatechin-3-monogallate PubChem CID: 65064 ChEBI: CHEBI:4806 IUPAC Name: [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate SMILES: C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
(-)-Epigallocatechin Gallate, MP Biomedicals™
CAS: 989-51-5 Molecular Formula: C22H18O11 Molecular Weight (g/mol): 458.375 InChI Key: WMBWREPUVVBILR-WIYYLYMNSA-N Synonym: =--epigallocatechin gallate,egcg,epigallocatechin gallate,epigallocatechin 3-gallate,tea catechin,epigallocatechin-3-gallate,=--epigallocatechin-3-o-gallate,teavigo,=--epigallocatechol gallate,epigallocatechin-3-monogallate PubChem CID: 65064 ChEBI: CHEBI:4806 IUPAC Name: [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate SMILES: C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
(-)-Epigallocatechin gallate, Thermo Scientific Chemicals
CAS: 989-51-5 Molecular Formula: C22H18O11 Molecular Weight (g/mol): 458.375 MDL Number: MFCD00075940 InChI Key: WMBWREPUVVBILR-WIYYLYMNSA-N Synonym: =--epigallocatechin gallate,egcg,epigallocatechin gallate,epigallocatechin 3-gallate,tea catechin,epigallocatechin-3-gallate,=--epigallocatechin-3-o-gallate,teavigo,=--epigallocatechol gallate,epigallocatechin-3-monogallate PubChem CID: 65064 ChEBI: CHEBI:4806 IUPAC Name: [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate SMILES: C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
MilliporeSigma™ Calbiochem™ (–)-Epigallocatechin Gallate
One of the main polyphenolic constituents of green tea that exhibits potent antitumor, anti-inflammatory, and antioxidant properties.
(-)-Epigallocatechin gallate, ≥98% (HPLC), MilliporeSigma™ Supelco™
(−)-Epigallocatechin gallate is a bioactive compound, which is an important constituent of green tea. It may serve as a potent candidate to prevent cancer.
Thermo Scientific Chemicals (-)-Epigallocatechin gallate, 95%
CAS: 989-51-5 Molecular Formula: C22H18O11 Molecular Weight (g/mol): 458.37 InChI Key: WMBWREPUVVBILR-WIYYLYMNSA-N Synonym: =--epigallocatechin gallate,egcg,epigallocatechin gallate,epigallocatechin 3-gallate,tea catechin,epigallocatechin-3-gallate,=--epigallocatechin-3-o-gallate,teavigo,=--epigallocatechol gallate,epigallocatechin-3-monogallate PubChem CID: 65064 ChEBI: CHEBI:4806 IUPAC Name: [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate SMILES: C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
(-)-Epigallocatechin Gallate Hydrate 98.0+%, TCI America™
CAS: 989-51-5 Molecular Formula: C22H18O11 Molecular Weight (g/mol): 458.375 MDL Number: MFCD00075940 InChI Key: WMBWREPUVVBILR-WIYYLYMNSA-N Synonym: =--epigallocatechin gallate,egcg,epigallocatechin gallate,epigallocatechin 3-gallate,tea catechin,epigallocatechin-3-gallate,=--epigallocatechin-3-o-gallate,teavigo,=--epigallocatechol gallate,epigallocatechin-3-monogallate PubChem CID: 65064 ChEBI: CHEBI:4806 IUPAC Name: [(2R,3R)-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate SMILES: C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O
R&D Systems™ EGCG
CAS: 989-51-5 Molecular Formula: C22H18O11
Epigallocatechin Gallate, G-Biosciences
CAS: 989-51-5 Molecular Weight (g/mol): 458.37 g/mol