Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (-)-Epinephrine (+)-bitartrate salt, 98+%

Catalog No. p-4229198
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430140010 1 g Glass bottle
AC430140050 5 g Glass bottle
AC430140250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC430140010 Supplier Thermo Scientific Chemicals Supplier No. 430140010
Only null left
Add to Cart

(-)-Epinephrine (+)-bitartrate salt, CAS # 51-42-3, also known as adrenaline, is a monoamine and catecholamine hormone.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • (-)-Epinephrine (+)-bitartrate salt, is a hormone
  • It is a monoamine and catecholamine

Applications

  • (-)-Epinephrine (+)-bitartrate salt is α-adrenergic and β-adrenergic receptor agonist
  • It can be used in adipose region to minimize blood loss
  • It is potentially a platelet agonist in preparation of optimal platelet aggregometry 96-well plates
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-42-3
Molecular Formula C13H19NO9
Molecular Weight (g/mol) 333.29
MDL Number MFCD00035077
InChI Key YLXIPWWIOISBDD-NDAAPVSOSA-N
Synonym L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate
PubChem CID 124080922
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 155.0°C
Color White to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,3674
Specific Rotation -20° to -24° (20°C, 589nm) (c=4, 1M HCl)
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Formula Weight 333.29
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material (-)-Epinephrine (+)-bitartrate salt
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Fatal if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye prot
EINECSNumber 200-097-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.