Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epinephrine Bitartrate, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18602419 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-419 5 g Amber Glass Bottle
18-602-420 25 g Amber Glass Bottle
18-602-422 100 g Amber Glass Bottle
18-602-421 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-602-419 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. EP1355GM
Add to Cart
Add to Cart

EP135, 51-42-3, C9H13NO3·C4H6O6

Spectrum™ Chemical Epinephrine Bitartrate, USP is used to treat life-threatening allergic reactions including anaphylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 51-42-3
Molecular Formula C13H19NO9
Molecular Weight (g/mol) 333.29
MDL Number MFCD00035077
InChI Key YLXIPWWIOISBDD-NDAAPVSOSA-N
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid; 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)C1=CC=C(O)C(O)=C1

Specifications

CAS Max % 100%
Ignition Residue Negligible
Loss on Drying 0.5%
Melting Range 147-152°C
Packaging Amber Glass Bottle
Quantity 5 g
UN Number UN2811
Percent Purity 97 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.