Learn More
Med Vet International Epinephrine inj, 1:1000 Multi Dose Vial, Vet Use
Supplier:  Med Vet International RXVEPI150
Encompass_Preferred
Multi-dose Vial. Non-refrigerated. A powerful, quick-acting vasoconstrictor for emergency use only in the treatment of anaphylactic shock in cattle, horses, sheep and swine. Administer subcutaneously or intramuscularly. Contains 1mg/mL epinephrine.
Catalog No. 50-282-8368
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.