Learn More
Epigentek Inc EpiQuik m6A RNA Methylation Quantification Kit (Colorimetric) (48 reactions)
Supplier: Epigentek Inc P900548
For absorbance-based quantitation of N6-methyladenosine in RNA in an ELISA-like format
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.