Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPLIN Antibody [DyLight 350], R&D Systems™
SDP

Catalog No. NBP327896UV
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP327896UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP327896UV Supplier R&D Systems Supplier No. NBP327896UV

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

EPLIN Polyclonal specifically detects EPLIN in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen EPLIN
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias 1110021C24Rik, Epithelial protein lost in neoplasm, EPLINepithelial protein lost in neoplasm beta, FLJ38853, LIM domain and actin binding 1, MGC131726, SREBP3LIM domain and actin-binding protein 1, sterol regulatory element binding protein 3
Host Species Rabbit
Immunogen EPLIN antibody was raised against an 18 amino acid peptide near the amino terminus of human EPLIN. The immunogen is located within amino acids 230 - 280 of EPLIN.
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51474
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.