Learn More
EPLIN Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
Specifications
Specifications
| Antigen | EPLIN |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | LIMA1 |
| Gene Accession No. | Q9UHB6 |
| Gene Alias | EPLIN, SREBP3, 1110021C24Rik, LIM domain and actin-binding protein 1, epithelial protein lost in neoplasm beta, sterol regulatory element binding protein 3 |
| Gene Symbols | LIMA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.