Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ EPLIN Recombinant Protein

Catalog No. 89012890 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-012-890 10 μg
89-012-891 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-012-890 Supplier Abnova™ Supplier No. H00051474P0110
Add to Cart
Add to Cart

Human EPLIN full-length ORF recombinant protein with GST-tag at N-terminal

EPLIN is a cytoskeleton-associated protein that inhibits actin filament depolymerization and cross-links filaments in bundles.

  • Theoretical MW (kDa): 91.52
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MENCLGESRHEVEKSEISENTDASGKIEKYNVPLNRLKMMFEKGEPTQTKILRAQSRSASGRKISENSYSLDDLEIGPGQLSSSTFDSEKNESRRNLELPRLSETSIKDRMAKYQAAVSKQSSSTNYTNELKASGGEIKIHKMEQKENVPPGPEVCITHQEGEKISANENSLAVRSTPAEDDSRDSQVKSEVQQPVHPKPLSPDSRASSLSESSPPKAMKFQAPARETCVECQKTVYPMERLLANQQVFHISCFRCSYCNNKLSLGTYASLHGRIYCKPHFNQLFKSKGNYDEGFGHRPHKDLWASKNENEEILERPAQLANARETPHSPGVEDAPIAKVGVLAASMEAKASSQQEKEDKPAETKKLRIAWPPPTELGSSGSALEEGIKMSKPKWPPEDEISKPEVPEDVDLDLKKLRRSSSLKERSRPFTVAASFQSTSVKSPKTVSPPIRKGWSMSEQSEESVGGRVAERKQVENAKASKKNGNVGKTTWQNKESKGETGKRSKEGHSLEMENENLVENGADSDEDDNSFLKQQSPQEPKSLNWSSFVDNTFAEEFTTQNQKSQDVELWEGEVVKELSVEEQIKRNRYYDEDEDEE

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH01247
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 51474
Molecular Weight (g/mol) 91.52
Name EPLIN (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MENCLGESRHEVEKSEISENTDASGKIEKYNVPLNRLKMMFEKGEPTQTKILRAQSRSASGRKISENSYSLDDLEIGPGQLSSSTFDSEKNESRRNLELPRLSETSIKDRMAKYQAAVSKQSSSTNYTNELKASGGEIKIHKMEQKENVPPGPEVCITHQEGEKISANENSLAVRSTPAEDDSRDSQVKSEVQQPVHPKPLSPDSRASSLSESSPPKAMKFQAPARETCVECQKTVYPMERLLANQQVFHISCFRCSYCNNKLSLGTYASLHGRIYCKPHFNQLFKSKGNYDEGFGHRPHKDLWASKNENEEILERPAQLANARETPHSPGVEDAPIAKVGVLAASMEAKASSQQEKEDKPAETKKLRIAWPPPTELGSSGSALEEGIKMSKPKWPPEDEISKPEVPEDVDLDLKKLRRSSSLKERSRPFTVAASFQSTSVKSPKTVSPPIRKGWSMSEQSEESVGGRVAERKQVENAKASKKNGNVGKTTWQNKESKGETGKRSKEGHSLEMENENLVENGADSDEDDNSFLKQQSPQEPKSLNWSSFVDNTFAEEFTTQNQKSQDVELWEGEVVKELSVEEQIKRNRYYDEDEDEE
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias EPLIN/FLJ38853/MGC131726/SREBP3
Common Name LIMA1
Gene Symbol LIMA1
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.