Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech Epo Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P31850UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P31850UG 50 μg
500P3181MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 500P31850UG Supplier PeproTech Supplier No. 500P31850UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human EPO. Anti-Human EPO-specific antibody was purified by affinity chromatography employing an immobilized Human EPO matrix. Sandwich ELISA: To detect Human EPO by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human EPO (500-P318Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human EPO. Western Blot: To detect Human EPO by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human EPO is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epo
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene EPO
Gene Accession No. P01588
Gene Alias EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2
Gene Symbols EPO
Host Species Rabbit
Immunogen CHO cells-derived Recombinant Human EPO.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2056
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.