Learn More
Epoxy Technology Inc EPO-TEK® 302 Epoxy Resin 8oz
SDP
Supplier:  Epoxy Technology Inc 3028OZ
Encompass
This item has a minimum qty of 6 per supplier requirements.
Two component, room temperature curing, medical grade epoxy featuring very low viscosity and excellent optical-mechanical properties. Application Type: Circuit / Electronic Assembly, Fiber Optic, Non-Conductive, Optical Materials, PCB Level Materials
Catalog No. NC0783935
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.