Learn More
Apexbio Technology LLC Amyloid Beta-peptide (25-35) (human)(Synonyms: Amyloid β-peptide (25-35), Aβ (25-35), Beta-amyloid (25-35), Amyloid beta-protein fragment (25-35)), 10mg, CAS: 131602-53-4.
SDP
Supplier:  Apexbio Technology LLC A103910
Encompass_Preferred
Amyloid Beta-peptide (25-35) (human) (CAS 131602-53-4) represents a neurotoxic fragment derived from the human -amyloid peptide This 11-amino acid segment possesses high aggregation propensity conferring pronounced toxic effects on neuronal cells relative to longer amyloid sequences Mechanistically the peptide promotes neurodegeneration memory impairment and cognitive dysfunction as evidenced by animal models displaying impaired spatial learning and memory following intracerebroventricular administration This peptide segment appears naturally in Alzheimer s disease (AD) patient brains especially within neurons of the subiculum and entorhinal cortex as well as in muscle tissues from individuals with inclusion body myositis As a result A (25-35) serves as an extensively utilized research tool in Alzheimer s pathology studies
Catalog No. 50-000-60415
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.