Learn More
Medchemexpress LLC Epothilone F | 208518-52-9 | MFCD19982658 | 99.5% | 523.68 g/mol | C27H41NO7S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1270795MG
Encompass_Preferred
Epothilone F is a microtubule/tubulin-stabilizing research compound used in cellular and biochemical studies. It exhibits anti-tumor activity and inhibits proliferation in various cancer cell lines. Supplied at high reported purity and available in small milligram quantities for laboratory research.
  • Microtubule/tubulin stabilizer used in cytoskeletal research.
  • Demonstrated anti-tumor cellular activity in vitro.
  • Supplied with reported purity of 99.45%.
  • Available in small milligram package sizes suitable for assays.
  • Provided in solid and solution formats for flexible use.
Catalog No. 50-002-64094
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.