Learn More
Ubpbio Epoxomicin, 1MG
Supplier:  Ubpbio F14001
Encompass_Preferred
Epoxomicin, originally isolated from Actinomyces sp., is a cell-permeable, potent, selective and irreversible proteasome inhibitor. It inhibits chymotrypsin-like, trypsin-like and caspase-like activities of the proteasome.
Catalog No. 50-196-5094
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.