Learn More
Eppendorf™ 24-Place Fixed-Angle Rotor w/Aerosol-tight Lid
Catalog No. 13683521 Shop All Eppendorf North America Products
Change view
Click to view available options
Type:
Rotor Key
Catalog No. Type
13-683-521 Rotor Key
1 options
Catalog No. 13-683-521 Supplier Eppendorf™ Supplier No. 022667181
Includes
Rotor and aerosol containment lid
For Models 5415D/5415R Microcentrifuges
  • 24 × 1.5 to 2.0mL capacity
  • Supplied with metal aerosol-tight lid for vapor containment
  • Maximum speed/RCF: 13,200rpm/16,110 x g
  • Autoclavable to 121°C (250°F)
Order Info
Accepts smaller tubes with accessory adapters, sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Key
For Use With (Equipment) Eppendorf 5415D/5415R Microcentrifuge