Learn More
Eppendorf™ 30-Place Fixed-Angle Rotor with Cover for Discontinued Model 5415C Microcentrifuge
Catalog No. 054063 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
24 x 250/500 μL Tubes
Catalog No. Capacity
05-406-3 24 x 250/500 μL Tubes
1 options
Catalog No. 05-406-3 Supplier Eppendorf™ Supplier No. 022366769
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
30 × 250 to 400μL capacity. Rotor speeds up to 14,000rpm/16,000×G.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 24 x 250/500 μL Tubes