Learn More
Eppendorf™ 5804/5810 Series Adapters for 05-400-94 Rotor
Catalog No. 054072 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
05-400-94 Swing bucket rotor
Catalog No. For Use With (Equipment)
05-407-2 05-400-94 Swing bucket rotor
1 options
Catalog No. 05-407-2 Supplier Eppendorf™ Supplier No. 022637452
Add to Cart
Add to Cart
For 50 mL tubes
  • Max speed 4,500 × g / 5,000 rpm
  • Radius 16.1 cm
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 05-400-94 Swing bucket rotor