Learn More
Eppendorf™ 5804/5810 Series Fixed-Angle Rotor
GSA_VA
Catalog No. 022639911 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
48 x 1.5/2 mL
Catalog No. Capacity
02-263-9911 48 x 1.5/2 mL
1 options
Catalog No. 02-263-9911 Supplier Eppendorf™ Supplier No. 022639911
Add to Cart
Add to Cart
Eppendorf QuickLock™ rotor lid for fast lid locking
Fixed-angle rotor FA-45-48-11 for Centrifuges 5804/5810, with software upgrade package.
  • Standard rotor for 48 × 1.5 / 2.0mL tubes (max. 18,210×g/14,000rpm)
  • Aerosol-tight and Eppendorf QuickLock-sealing
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Fixed Angle Rotor
Capacity 48 x 1.5/2 mL
Max. RCF 18,210 x g
Max. Speed 14,000 rpm
For Use With (Equipment) 5804/5810 Centrifuge
Includes Software upgrade package, Aerosol-tight and QuickLock™ sealing