Learn More
Eppendorf™ 5804/5810 Series Tube Adapters for 05-400-81 Fixed Angle Rotor
Catalog No. p-3206387 Shop All Eppendorf North America Products
Change view
Click to view available options
Capacity:
15 mL Conical Tubes
50 mL Conical Tubes
50 mL Tubes
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
05-400-86 50 mL Tubes
05-400-87 50 mL Conical Tubes
05-406-30 15 mL Conical Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-400-86 Supplier Eppendorf™ Supplier No. 022637258
Only null left
Add to Cart
Add to Cart
For use with 05-400-81 Fixed Angle Rotor
Adapters are supplied in packs of two. Order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Capacity 50 mL Tubes
For Use With (Equipment) Fixed angle rotor