Learn More
Eppendorf™ 5810 Series Four-Place Swinging Microplate Buckets
GSA_VA
Catalog No. 0540079 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
A-4-62 MTP swing bucket rotor for 5810/5810R centrifuge
Catalog No. For Use With (Equipment)
05-400-79 A-4-62 MTP swing bucket rotor for 5810/5810R centrifuge
1 options
Catalog No. 05-400-79 Supplier Eppendorf™ Supplier No. 022638068
Add to Cart
Edge
Add to Cart
For use with 05-400-64 rotor to spin microplates
  • Each bucket accepts up to four standard microplates for a maximum of 16 microplates per run
  • Maximum rotor speed/force: 4000rpm/2750 x g
  • Buckets can also be used in 05-400-80 rotor
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) A-4-62 MTP swing bucket rotor for 5810/5810R centrifuge