Learn More
Eppendorf™ Lid for ThermoMixer™ F1.5 and FP
GSA_VA
Catalog No. 05412517 Shop All Eppendorf North America Products
Change view
Click to view available options
Type:
Plate Lid
Catalog No. Type
05-412-517 Plate Lid
1 options
Catalog No. 05-412-517 Supplier Eppendorf™ Supplier No. 5363000233
Add to Cart
Edge
Add to Cart
Use this lid with ThermoMixer™ F1.5 and SmartBlocks™ 0.5mL, 1.5mL, 2.0mL, plates PCR 96, PCR 384. Eppendorf™ Lid for ThermoMixer™ F1.5 and FP is exchangeable in seconds.
Maintain accurate, uniform temperatures with individually sensor-controlled SmartBlocks.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 0.5, 1.5, 2mL SmartBlocks, 96 and 384-well PCR plates
Type Plate Lid