Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Combitips™ Advanced Tips for Multipette™ Multi-dispensers - Eppendorf Quality
GSA_VAGREENER_CHOICE

Catalog No. 13683701 Shop All Eppendorf North America Products
Click to view available options
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
1.0 mL
2.5 mL
5.0 mL
10 mL
25 mL
50 mL
Color Code:
Blue
Green
Light Blue
Light Gray
Orange
Red
Violet
White
Yellow

Eppendorf's consumable of choice for all Eppendorf Multipette™ multi-dispensers. Available in 9 volume sizes (0.1 to 50 mL), Combitips advanced in 'Eppendorf Quality' dispense in increments as low as 100 mL and are ideally suited for a variety of standard lab applications. Also available in purity grades PCR clean, Biopur™ or Forensic DNA Grade.

Use these tips for comfortable handling while preventing damage to gloves. Eppendorf Combitips advanced™ Standard Pipetter Tips are optimized to meet the increased requirements of a modern laboratory, and offered in three different purity grades to support any application.

  • Combitips advanced are the ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 mL – 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • An individual color coding facilitates the quick identification of the desired Combitips

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.2 mL
Filtered Non-filtered
Sterility Non-sterile
Color Colorless
Autoclavable Not Autoclavable
Clean Claims Eppendorf Quality (Non-sterile)
Format Bulk, Bagged
Tip Style Extended Length, Positive Displacement
Packaging 100 Pk. (4 Bags x 25 Pk.)
Product Type Pipette Tips
Color Code Light Blue
Disposable Single-use
For Use With (Equipment) Positive Displacement System
Graduated Yes
Product Line CombiTips Advanced
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.