Learn More
Eppendorf™ Combitips™ Tips
Catalog No. 21381307E Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
12.5 mL
Catalog No. Volume (Metric)
21-381-307E 12.5 mL
1 options
This item is not returnable. View return policy
Catalog No. 21-381-307E Supplier Eppendorf™ Supplier No. 022495208
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
For Eppendorf Repeater™ Pipetter
Tips have polyethylene plunger and polypropylene cylinder, making them ideal for use with most reagents
Order Info
Nonsterile tips are packed in plastic bags. Biopur™ tips are individually wrapped.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 12.5 mL
Sterility Sterile
Tip Style Specialty
For Use With (Equipment) Eppendorf Repeater
Product Line CombiTips
Material Polypropylene Cylinder, Polyethylene Plunger