Learn More
Eppendorf 3P Eppendorf Conical Tubes 25 mL with SnapTec™ cap, 25 mL, Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, colorless, 150 tubes (6 bags × 25 tubes)
Supplier:  Eppendorf 3P 0030118421
Encompass_Preferred
Reduced height gives easy sample access even with low-volume pipettes and tips – reduced risk of cross-contamination between pipette and tube. Single-handed opening and closing through SnapTec™ cap version for fast liquid extraction and addition. Centrifugation stability up to 17,000 x g1) for both tube variants - SnapTec™ cap and screw cap. Tightness of the lid is ensured over a broad temperature range. Less storage space needed due to ~20 % lower height compared to 50 mL conical tubes. Matching accessories for centrifugation, mixing and heating, sample preparation and storage ensure straightforward integration into existing laboratory environment. Premium USP class 6 raw material and no use of slip agents, plasticizers, and biocides during manufacturing ensure highest sample integrity and optimal sample and pellet visibility. Screw cap with flattened and grooved sides supports a user-friendly ergonomic handling, enables a stable upright positioning.
Catalog No. NC1689261
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.