Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Eppendorf Totally Integrated Pipetting System Tips work in perfect combination with pipettes. Best results are obtained while pipetting from and into 15mL conical tubes, high reagent bottles, narrow deep vessels and especially Eppendorf Deepwell Plates.
- Extended length for working with narrow vessels
- Drop contour design gives all Eppendorf tips a uniform look
- Soft, flexible tip cone ensures better pipette tip sealing
- Ergonomically optimized cone geometry ensures perfect fit on Eppendorf pipettes
- Universal fit so can be used with pipettes from other manufacturers
- Fine graduations make it easy to visually check the volume pipetted
- New tip design for better access to a wider variety of tubes and wells
- Finely tapered and extended for working with small volumes
- Cross-contamination during pipetting can be avoided to a minimum
- Even smallest volumes in deepwell plates of 96 and 384 wells can be almost completely recovered from the bottom
Specifications
Specifications
| Volume (Metric) | 50 to 1250 μL |
| Color | Green |
| Length (English) | 4.05 in. |
| Length (Metric) | 10.3cm |
| For Use With (Application) | Reduced-force tip attachment and ejection with complete sealing |
| For Use With (Equipment) | Pipette |
| Product Line | epT.I.P.S. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?