Learn More
Eppendorf™ Geloader Pipet Tips
Catalog No. 213802m Shop All Eppendorf North America Products
Change view
Click to view available options
Pipet Tips, Case of 192
Catalog No.
21-380-2M Pipet Tips, Case of 192
1 options
Catalog No. 21-380-2M Supplier Eppendorf™ Supplier No. 022351656
Reliable, reproducible, high-precision pipetting for a wide variety of applications
  • Drop contour design
  • Finely tapered and extended; ideal for small-volume pipetting
  • Fine graduations make it easy to check volume
  • Soft, flexible tip cone provides better seal to pipetter
Certifications
ISO 9001:2000
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 20 μL
Color Clear
Certifications/Compliance ISO 9001:2000
Disposable Single-use
Graduated Yes
Material Virgin Polypropylene