Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Maxipettor™

Catalog No. 2137925 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
1 to 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-379-25 1 to 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-379-25 Supplier Eppendorf™ Supplier No. 022290002

missing translation for 'validMainframeMsg'

Digital volume display ensures exact, reproducible results

  • Select volumes from 1 to 10mL, in 0.01mL increments
  • Adjustable from 1 to 10mL
  • Accuracy to ±0.3%; precision to 0.1%
  • Replaces conventional transfer and serological pipets
  • Can be used for aqueous and organic solutions, as well as liquids with high vapor pressure, density or viscosity
  • Disposable tips ensure safety and accuracy by eliminating carryover
  • Reagents contact the tip only— never the Maxipettor unit itself

Order Info

Two types of tips are available: air displacement (Maxitip S™ and Maxitip G™ tips) and positive displacement (Maxitip P™ tips).

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.3%
Autoclavable Not Autoclavable
Number of Channels 1
Type Adjustable Pipette
Volume (Metric) 1 to 10 mL
For Use With (Application) Aqueous and organic solutions, as well as liquids with high vapor pressure, density or viscosity
Increments 10μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.