Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Model 4830 Positive Displacement Pipette Tips/Pistons

Catalog No. 2137972 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-379-72 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-379-72 Supplier Eppendorf™ Supplier No. 022354159

missing translation for 'validMainframeMsg'

Eppendorf positive displacement tips have been optimized for handling viscous liquids, high vapor pressure liquids or precious samples. The piston is contained within the tip, allowing for complete dispensing of liquids. Positive displacement tips are for use with the BioMaster positive displacement pipette.

  • Polypropylene tips; polyethylene pistons
  • Mounted ready for use
  • In five autoclavable racks of 96 tips/pistons

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Color Light Gray
Autoclavable Autoclavable
Format Rack
Tip Style Positive Displacement
For Use With (Equipment) BioMaster Positive Displacement Pipette
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.