Learn More
Eppendorf™ Repeater™ Pipetter, Combitube™ Accessory
Catalog No. 213802A Shop All Eppendorf North America Products
Change view
Click to view available options
Type:
Aspirating Aid
Catalog No. Type
21-380-2A Aspirating Aid
1 options
Catalog No. 21-380-2A Supplier Eppendorf™ Supplier No. 022261550
Provides a safe and convenient way to fill Eppendorf™ Combitips™ tips directly from tall, narrow-neck bottles; eliminates need to transfer liquids into secondary vessel. Consists of circular liquid reservoir, valve, and long filling tube. Reagent supply in reservoir is constantly replenished as liquid is drawn out. Made of chemical-resistant polypropylene; autoclavable.
TRUSTED_SUSTAINABILITY
Specifications
Type Aspirating Aid
For Use With (Equipment) 5, 25 and 50mL Repeater™ pipette
Functional Range 5, 25 and 50 mL