Learn More
Sigma Aldrich Fine Chemicals Biosciences Eppendorf ResearchR Plus G1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP31230000201EA
Encompass
The Eppendorf Research plus mechanical pipette combines over 50 years of innovation in liquid handling and is one of the most commonly used pipettes in the world. It provides you with one of the safest and most ergonomic pipettes available today. The Research plus microliter pipette protects the health of our customers during their daily work in the laboratory. The Research plus is the pipette family with the lowest weight and lowest operation forces in the Eppendorf product families. Lower operation forces can only be reached with our Eppendorf electronic pipettes. Enjoy excellent flexibility and choose among single-channel pipettes in fixed or variable volume as well as multi-channel pipette options with 8 12 16 and 24 channels.Product featuresIt is a good feeling to know you are doing the right thing - to know you are working with one of the most advanced pipettes in the world. A spring loaded tip cone a secondary calibration option an improved volume display - and all that in a
Catalog No. 50-290-4303
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.