Learn More
Eppendorf™ Rotor Spacer for Vacufuge™ Concentrator
Catalog No. 0774822 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Vacufuge™ Concentrator
Catalog No. For Use With (Equipment)
07-748-22 Vacufuge™ Concentrator
1 options
Catalog No. 07-748-22 Supplier Eppendorf™ Supplier No. 022822101
Add to Cart
Add to Cart
Rotor for Vacufuge™ Concentrator may be stacked for doubled capacity.
  • Rotors are designed for microcentrifuge tubes; may be stacked for doubled capacity in Eppendorf™ Vacufuge™ Concentrator.
1 Required when using two stackable rotors during a single run.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Spacer
For Use With (Equipment) Vacufuge™ Concentrator
Temperature (Metric) Operating 30, 40, 65°C
Voltage 115 V
Frequency 50/60 Hz