Learn More
Eppendorf™ Stacking Kit for Innova 44/44R Incubator Shaker
Catalog No. 13683525 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Innova 44/44R Incubator Shaker
Catalog No. For Use With (Equipment)
13-683-525 Innova 44/44R Incubator Shaker
1 options
Catalog No. 13-683-525 Supplier Eppendorf™ Supplier No. M12820500
Add to Cart
Edge
Add to Cart
Provides additional stacking capability
  • For stacking one additional 44 or 44R Shaker
  • Bolts together shakers for secure mounting
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stacking Kit
For Use With (Equipment) Innova 44/44R Incubator Shaker