Learn More
Eppendorf™ ThermoMixer™ C Accessory, Smartblock™ Thermoblock
GSA_VA
Catalog No. 05412512 Shop All Eppendorf North America Products
Change view
Click to view available options
Holds:
96-well PCR Plate
Catalog No. Holds
05-412-512 96-well PCR Plate
1 options
Catalog No. 05-412-512 Supplier Eppendorf™ Supplier No. 5306000006
Add to Cart
Add to Cart
Maintain accurate uniform temperatures with these thermoblocks. Eppendorf™ Smartblock™ Thermoblocks are compatible with Eppendorf ThermoMixer™ C, and support a broad range of applications.
  • Exchangeable in just seconds
  • Automatic block recognition for safe handling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Eppendorf ThermoMixer C
Holds 96-well PCR Plate
Type Block
Wash/Waste Bottle Capacity 96-well PCR Plate