Learn More
Eppendorf™ Thermomixer™ R Accessory, Thermoblock Dry Blocks
Catalog No. 0540035 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Application):
For one PCR and deepwell plate
Catalog No. For Use With (Application)
05-400-35 For one PCR and deepwell plate
1 options
Catalog No. 05-400-35 Supplier Eppendorf™ Supplier No. 022670565
These exchangeable thermoblocks for the Thermomixer R and ThermoStat provide excellent flexibility. The range of potential applications is significantly expanded due to the support for slides and microarrays, in addition to a broad range of tube sizes.
  • Shielded thermoblocks are contact-safe
  • Efficient exchange simplifies operation
  • Tubes are visible during operation and easily accessible
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For one PCR and deepwell plate
Temperature Range (Metric) 70°C
Type Block
For Use With (Application) For one PCR and deepwell plate
Includes Lid for improved temperature accuracy
Speed 1400rpm
Height (English) 4.9 in.
Height (Metric) 124 mm