Learn More
Eppendorf™ Tube Adapters for 36-Place Fixed-Angle Rotor
Catalog No. 05400128 Shop All Eppendorf North America Products
Change view
Click to view available options
Bore Size (Metric):
0.6 cm
Catalog No. Bore Size (Metric)
05-400-128 0.6 cm
1 options
Catalog No. 05-400-128 Supplier Eppendorf™ Supplier No. 022636286
Add to Cart
Add to Cart
For use with 5415D/5415R Microcentrifuges
Fit directly in 05-400-127 rotor to accommodate 0.2mL PCR tubes.
Order Info
Adapters are supplied in packs of six. Order the proper quantity to fully equip the rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Eppendorf special rotor for 5415D or 5415R microcentrifuge
Bore Size (Metric) 0.6 cm