Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Tube Clip for Tubes 5mL
GREENER_CHOICE

Catalog No. 14282322 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Eppendorf tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-282-322 Eppendorf tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-282-322 Supplier Eppendorf™ Supplier No. 0030119509
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Securely close tubes of 5.0mL with these sturdy clips.

  • Supplied in packs of 10
  • Material: Exceptionally high-quality, transparent polypropylene, free of plasticizers, biocides or mold release agents, for reliable test results
  • Certified purity: Available in lot specific certified Eppendorf Quality, PCR clean, Sterile and Eppendorf Biopur™ purity grades.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Tube Clip
For Use With (Equipment) Eppendorf tubes
Packaging Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.