Learn More
Sigma Aldrich Fine Chemicals Biosciences Eppendorf TubeR capacity 5
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0030119487200EA
Encompass
Eppendorf Tubes are the perfect option for working with medium-sized sample volumesMaximum safety and stability for centrifugation up to 25 000 x gConical bottomsEasier processing of samples up to 5.0 mLHigher yields in DNA isolation prep of mastermixes and buffers and safe cell and tissue lysisTransparent polypropylene free of plasticizers biocides or mold release agentsHinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86C to 80C
Catalog No. 50-290-1086
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.