Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ PCR Plates 384-well, 45 μL - PCR Plates
GREENER_CHOICE

Catalog No. E0030129342 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Blue
Colorless
Green
Red
Yellow
Barcode:
Alphanumeric
No Barcode
Packaging:
10 Bags x 30 Plates
12 Bags x 25 Plates
5 Bags x 5 Plates
Individually Blister Wrapped
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Barcode Packaging Purity or Quality Grade
E0030129342 Colorless No Barcode Individually Blister Wrapped PCR Clean (Non-sterile)
E0030129350 Blue No Barcode Individually Blister Wrapped PCR Clean (Non-sterile)
E0030129628 Colorless No Barcode Individually Blister Wrapped Forensic DNA Grade (Non-sterile)
E951020737 Blue No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020711 Yellow No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020729 Green No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020745 Red No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020702 Colorless No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
14-380-055 Colorless No Barcode 5 Bags x 5 Plates PCR Clean (Non-sterile)
E951020539 Colorless No Barcode 10 Bags x 30 Plates PCR Clean (Non-sterile)
E951020573 Blue No Barcode 10 Bags x 30 Plates PCR Clean (Non-sterile)
01-671-102 Colorless Alphanumeric 5 Bags x 5 Plates PCR Clean (Non-sterile)
14-225-010 Red No Barcode 12 Bags x 25 Plates PCR Clean (Non-sterile)
14-225-009 Green No Barcode 12 Bags x 25 Plates PCR Clean (Non-sterile)
14-225-008 Yellow No Barcode 12 Bags x 25 Plates PCR Clean (Non-sterile)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. E0030129342 Supplier Eppendorf™ Supplier No. 0030129342
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean).
  • Safe Code option provides 3-level coding for safe sample and vessel identification.
  • ECC 200 error correction function enables safe code reading with up to 30% damaged codes.

Certifications

Certified free of human DNA, DNase, RNase, and PCR inhibitors

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Free of Human DNA, DNase, RNase and PCR Inhibitors
Format Standard
Array 16 x 24
Autoclavable Autoclavable
Packaging Individually Blister Wrapped
Color Colorless
Product Type PCR Plate
For Use With (Equipment) Automated Systems
Skirt Style skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 45 μL
No. of Wells 384
Sterility Non-sterile
Well Shape Round Bottom (U-well)
Max. RCF 2250 x g
Purity or Quality Grade PCR Clean (Non-sterile)
Quantity 10 Pk.
Well Color Colorless
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.