Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ twin.tec™ Real-Time 96-Well PCR Platesc

Catalog No. 14281213 Shop All Eppendorf North America Products
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-281-213 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-281-213 Supplier Eppendorf™ Supplier No. 0030132750

missing translation for 'validMainframeMsg'

Fluorescence intensity up to ten times higher than with frosted wells

The limiting factor in low volume real-time PCR is often the remaining intensity of fluorescence. The white wells of the Eppendorf twin.tec real-time PCR plates reflect fluorescence much better than clear or frosted wells. Thus, lower levels of fluorescence are still detectable with the same instrument–just by using the right consumables. Additionally, white wells significantly reduce interfering background fluorescence and lead to increased homogeneity of replicates and reproducible results.

  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121°C, 20 min.)
  • Rigid polycarbonate frame ensures easy and reliable handling, manually or automated
  • Polypropylene wells guarantee excellent and fast temperature transfer to the sample

TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Array 8 x 12
Autoclavable Autoclavable
Color Black
Material Polypropylene
No. of Wells 96
Sterility Sterile
For Use With (Application) RT-PCR applications
Rimmed Yes
Well Color White
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.