Learn More
Sigma Aldrich Fine Chemicals Biosciences Eppendorf twin.tecR PCR Pl
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP003012951225EA
Encompass
Eppendorf twin.tec(R) 96 Well PCR Plates improve recovery of nucleic acids by reducing their absorption to the tube wall. A combination of special manufacturing technologies and selected polypropylene batches ensures nearly 100 % recovery of DNA/RNA molecules without surface coating to eliminate the risk of sample contamination.Batch-tested and certified by an independent laboratory to be free from DNA DNase RNase and PCR inhibitors.
Catalog No. 50-290-0712
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.