Learn More
Eppendorf™ Vis Cuvettes
Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm. The Vis cuvettes are the ideal tool for applications outside of the UV range, for example, colorimetric protein assays (Bradford, Lowry, etc.), determining of the optical density of bacterial cultures (OD 600 methods), kinetic, and fluorescence measurements.
$371.11 - $395.89
Specifications
Material Plastic
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
14-281-215
View Documents
Eppendorf™
0030079353
Semi-micro Vis Cuvet
Case of 1000 for $395.89
Only null left
Add to Cart
14-281-216
View Documents
Eppendorf™
0030079345
Macro Vis Cuvet
Case of 1000 for $371.11
Only null left
Add to Cart
Description
Description
  • Two different cuvette sizes (semi-micro and macro) for a wide variety of volumes and applications
  • Disposable cuvettes made of clear plastic with a light transmission of 300 nm to 900 nm
Specifications
Specifications
Plastic
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.