Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Centrifuge 54181EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP54010001371EA
Encompass
The Centrifuge 5418 and Centrifuge 5418 R with their 18-place capacity rotors are specifically designed for low to medium capacity in 1.5/2.0 mL tubes and microtainer. Their speed of up to 16873 x g allows for standard molecular biology and clinical applications. Both models have an especially low access height and are characterized by a remarkably quiet operation.
Catalog No. 50-290-8965
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.