Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Centrifuge 54251EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP54050000421EA
Encompass
Centrifuge 5425 is the new laboratory standard with its 24-place capacity and speed up to 21 330 x G. Focusing on standard applications it is an essential piece of equipment for modern labs ensuring optimal separation. The centrifuge 5425 serves as a reliable Partner especially for high-throughput users Expecting a top level of ergonomic comfort and provides great flexibility with its 6 rotor options.
Catalog No. 50-290-9039
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.