Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Centrifuge 54271EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0226207001EA
Encompass
The Centrifuge 5427 R was designed with high-throughput research applications in mind. With its aerosol-tight 48-place rotor it handles even the toughest challenges in a safe and reliable manner to provide you the efficiency boost you need. All aerosol-tight rotors including the new rotors for Eppendorf Tube(R) 5.0 mL as well as the 24 x 1.5/2.0 mL swing-bucket rotor are equipped with the Eppendorf QuickLock(R) lids for easy rotor lid opening and closing. Choose the Centrifuge 5427 R if a 24-place microcentrifuge (e.g. our Centrifuge 5424 R) doesnt meet your needs regarding tube capacity and if you dont need the extra versatility our cross-over Centrifuge 5430 R can offer you.Soft-touch lid closureCentrifuge 5427 R is equipped with a soft--touch one-finger lid closure for ergonomic operation. Just lower the lid gently and it will lock automatically. No more pressing or slamming down the lid.
Catalog No. 50-290-9052
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.