Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Centrifuge 54301EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0226205841EA
Encompass
Unique cross-over Centrifuge 5430 and 5430 R combine the best features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument. These centrifuges spin rotors for TMEppendorf tube] and PCR strips as you would expect from any microcentrifuge. But thats not all. In a compact size Centrifuge 5430 and 5430 R also accommodates rotors for microplates and 15/50 mL conical tubes. Now the rotor program has been expanded with additional Eppendorf QuickLock(R) rotors. This includes 48-place rotors (1.5/2.0 mL) a 16-place rotor for Eppendorf Tube(R) 5.0 mL and a swing-bucket rotor (24 x 1.5/2.0 mL). With their unmatched versatility Centrifuge 5430 and Centrifuge 5430 R are the flagship products among the Eppendorf microcentrifuge portfolio. Available as keypad or knob variant
Catalog No. 50-290-8953
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.