Learn More
Sigma Aldrich Fine Chemicals Biosciences EppendorfR Centrifuge 57021EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences EP0226262051EA
Encompass
The 5702 family of low speed general purpose centrifuges was especially designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs. They are compact in design so that one can fit comfortably on every lab bench quiet in operation to benefit your work environment and provide 6 different rotor options to accommodate any type of tube.The right model for your application.nbspCentrifuge 5702 for standard applications .nbspRefrigerated Centrifuge 5702 R for heat sensitive samples .nbspHeated and refrigerated Centrifuge 5702 RH allows centrifugation under biological conditions to improve cell viability for more accurate results in downstream applications (e.g. cell cultivation)Ergonomics.nbspVery compact footprint to fit on every lab bench .nbspVery quiet operation to benefit your environment .nbspLow access height for simple loading and unloading of samples Temperature management.nbspOptimizes air flow minimizes sample warming during run (5702) .nbsp
Catalog No. 50-290-8825
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.